EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Enantioselective total synthesis of hyperforin.
PMID 23270309
Abstract
A modular, 18-step total synthesis of hyperforin is described. The natural product was quickly accessed using latent symmetry elements, whereby a group-selective, Lewis acid-catalyzed epoxide-opening cascade cyclization was used to furnish the bicyclo[3.3.1]nonane core and set two key quaternary stereocenters.