EMAIL THIS PAGE TO A FRIEND
BMC Cell Biology
Pressure mediated hypertrophy and mechanical stretch up-regulate expression of the long form of leptin receptor (ob-Rb) in rat cardiac myocytes.
PMID 23270329