EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
Acute toxicity of cypermethrin to the non target organism Hyalella curvispina.
PMID 23270860
Abstract
The acute toxicity of cypermethrin to the amphipod Hyalella curvispina was evaluated by means of a toxicity test under laboratory conditions. Cypermethrin is one of the most widely used insecticides in Argentina. H. curvispina is a widely distributed and commonly abundant component of the invertebrate assemblages in shallow waters of southern South America. The experiments were repeated three times. The mean 48-h LC(50) value for H. curvispina was estimated at 0.066μg/l. H. curvispina represents a good model for exotoxicological risk assessment.