EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Mucosa-plate for direct evaluation of mucoadhesion of drug carriers.
PMID 23270997
Abstract
The method to prepare mucosa-plates, glass slides covalently coated with mucin, is demonstrated. The use of the plate to evaluate mucoadhesion of nanocarriers made from different four polymeric materials, N-succinylchitosan (NS-chitosan), alginate (ALG), ethylcellulose (EC), and a blend of EC and methylcellulose (EC/MC), was demonstrated. While different mucoadhesion of the four carriers could be detected using mucosa-plate, the conventional viscosity measurement could not differentiate their mucin-binding ability. ALG and NS-chitosan nanospheres showed the best attachment to the mucosa-plate compared to the EC/MC and EC spheres. Capsaicin, a model hydrophobic drug, was loaded into the carriers and the ability of the different polymeric carriers to retain capsaicin at the stomach tissue was compared using an ex vivo fresh porcine stomach assay. Ability to retain capsaicin at the stomach tissue correlated well with binding affinity toward the mucosa-plate and the loading capacity of the carriers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
200646
Ethyl cellulose, viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
200689
Ethyl cellulose, viscosity 10 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
200697
Ethyl cellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
433837
Ethyl cellulose, viscosity 46 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
247499
Ethyl cellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
200654
Ethyl cellulose, viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
46070
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis
46080
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis
64655
Methocel® 60 HG, 28-30% methoxyl basis
64670
Methocel® 65 HG, 27-29% methoxyl basis
64680
Methocel® 90 HG, 22-27% methoxyl basis
64605
Methocel® A15 LV, 27.5-31.5% methoxyl basis
64625
Methocel® A15C, medium viscosity, Methoxyl content 27.5-31.5 %
94378
Methocel® A4M, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
M0262
Methyl cellulose, viscosity: 400 cP
M7140
Methyl cellulose, viscosity: 15 cP
M6385
Methyl cellulose, viscosity: 25 cP
M7027
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
M0387
Methyl cellulose, viscosity: 1,500 cP
M0430
Methyl cellulose, meets USP testing specifications, viscosity: 400 cP
M0555
Methyl cellulose, meets USP testing specifications, viscosity: 1,500 cP
64632
Methyl cellulose, tested according to Ph.Eur.