EMAIL THIS PAGE TO A FRIEND
Atherosclerosis
Angiotensinogen gene polymorphism and HDL2 are linked to coronary artery calcification in individuals with family history of early coronary disease.
PMID 23273503