EMAIL THIS PAGE TO A FRIEND
Veterinary parasitology
Identification of a field isolate of Fasciola hepatica resistant to albendazole and susceptible to triclabendazole.
PMID 23273779
Abstract
The experiments described here were designed to characterize the status of susceptibility/resistance to albendazole (ABZ) and triclabendazole (TCBZ) of a Fasciola hepatica isolate (named CEDIVE isolate) recovered from infected sheep (Gualeguay, Argentina) and maintained under laboratory conditions. Two separate clinical efficacy experiments were performed on sheep artificially infected with the CEDIVE isolate. Experiment 1: sheep were randomly distributed either in an untreated control group or in an ABZ (7.5 mg/kg) treated group (n=4 each). Additionally, the systemic exposure of ABZ metabolites was assessed in those ABZ-treated infected animals. In Experiment 2, an untreated control group and a TCBZ (10 mg/kg) treated group was included (n=4 each). The fluckicidal efficacy of ABZ and TCBZ was assessed by the comparison of the number of flukes recovered from untreated and treated sheep at 15 days post-treatment. The efficacy against the CEDIVE isolate of F. hepatica was 29% (ABZ) and 100% (TCBZ). The plasma drug exposure (expressed as AUC and C(max)) observed in the ABZ treated animals (Experiment 1) was in agreement with data obtained in the previous studies, which indicate that the low ABZ efficacy was not related to the quality of the pharmaceutical product and/or to a low systemic availability of the active drug/metabolite. The results reported here clearly show that the CEDIVE isolate of F. hepatica behaves as resistant to ABZ and susceptible to TCBZ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32802
Triclabendazole, VETRANAL, analytical standard
C14H9Cl3N2OS