EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Submerged-arc welding slags: characterization and leaching strategies for the removal of aluminum and titanium.
PMID 23274794
Abstract
In the present study, submerged-arc welding slags were characterized by applying a variety of methods, including X-ray fluorescence, X-ray diffraction, particle size, Raman spectroscopy, and scanning electron microscope with energy dispersive X-ray analysis. The content of Al proved to be quite similar within neutral and acid slags (10-14%), while that of Ti proved to be much higher in acid slags (approximately 10%) than in neutral slags (<1%). The presence of spinel structures associated with Al species could also be identified in the analyzed samples. This characterization study was accompanied by leaching tests performed under changing operating conditions in an attempt to evaluate to what extent the Al and Ti bearing components could be removed from the slags. The leaching work involved three distinct strategies: (i) NaOH leaching followed by H(2)SO(4) leaching, (ii) acid leaching (HCl and H(2)SO(4)) using oxidizing/reducing agents, and (iii) slag calcination followed by H(2)SO(4) leaching. In the best result, 80% of Al was extracted in one single leaching stage after calcination of the acid slag with NaCl+C at 900 °C. By contrast, the removal of Ti proved to be unsatisfactory.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223913 Ascarite®, Sodium hydroxide-coated silica, 8-20 mesh
223921 Ascarite®, Sodium hydroxide-coated silica, 20-30 mesh
S8045
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
HNaO
306576
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
HNaO
S5881
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
HNaO
484024
Sodium hydroxide, reagent grade, 97%, flakes
HNaO
30620
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
HNaO
06203
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
HNaO
06306
Sodium hydroxide, ≥98.5%, microprills
HNaO
06208
Sodium hydroxide, for the manufacture of mirrors, pellets, Cl <0.001%
HNaO
655104
Sodium hydroxide, reagent grade, 97%, powder
HNaO
901915
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
HNaO
415413
Sodium hydroxide solution, 50% in H2O
HNaO
05211
Sodium hydroxide solution, purum, ≥32%
HNaO
30530
Sodium hydroxide solution, ≥20%
HNaO
S8263
Sodium hydroxide solution, 5.0 M
HNaO
320501
Sulfuric acid, ACS reagent, 95.0-98.0%
H2O4S
258105
Sulfuric acid, ACS reagent, 95.0-98.0%
H2O4S
30743
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
H2O4S
07208
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
H2O4S
84736
Sulfuric acid solution, puriss. p.a., ≥25% (T)
H2O4S