EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Changes in pH and organic acids in mucilage of Eriophorum angustifolium roots after exposure to elevated concentrations of toxic elements.
PMID 23274805
Abstract
The presence of Eriophorum angustifolium in mine tailings of pyrite maintains a neutral pH, despite weathering, thus lowering the release of toxic elements into acid mine drainage water. We investigated if the presence of slightly elevated levels of free toxic elements triggers the plant rhizosphere to change the pH towards neutral by increasing organic acid contents. Plants were treated with a combination of As, Pb, Cu, Cd, and Zn at different concentrations in nutrient medium and in soil in a rhizobox-like system for 48-120 h. The pH and organic acids were detected in the mucilage dissolved from root surface, reflecting the rhizospheric solution. Also the pH of root-cell apoplasm was investigated. Both apoplasmic and mucilage pH increased and the concentrations of organic acids enhanced in the mucilage with slightly elevated levels of toxic elements. When organic acids concentration was high, also the pH was high. Thus, efflux of organic acids from the roots of E. angustifolium may induce rhizosphere basification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
156264
Ammonium formate, reagent grade, 97%
CH5NO2
713716
Basosiv M050, produced by BASF
C2H2MgO4
21134
Calcium formate, BioUltra, ≥99.0% (T)
C2H2CaO4
03826
Calcium formate, Standard for quantitative NMR, TraceCERT®
C2H2CaO4
433764
Cesium formate, 98%
CHCsO2
46940-U
D-Malic acid, analytical standard
C4H6O5
02300
D-(+)-Malic acid, unnatural form, ≥97.0% (T)
C4H6O5
240176
DL-Malic acid, ReagentPlus®, ≥99%
C4H6O5
M0875
DL-Malic acid, ≥98% (capillary GC)
C4H6O5
W265501
DL-Malic acid, 99%
C4H6O5
27606
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
C4H6O5
M6773
DL-Malic acid disodium salt, ≥95% (capillary GC)
C4H4Na2O5
F0507
Formic acid, reagent grade, ≥95%
CH2O2
33015
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
CH2O2
695076
Formic acid, ACS reagent, ≥96%
CH2O2
27001
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
CH2O2
399388
Formic acid, ACS reagent, 88-91%
CH2O2
M9138
L-(−)-Malic acid disodium salt, 95-100% (enzymatic)
C4H4Na2O5
M8304
Malic acid, meets USP/NF testing specifications
C4H6O5
68487
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
C2H2O4
O0130
Oxalic acid dilithium salt
C2Li2O4
294454 Potassium formate, ReagentPlus®, 99%
CHKO2
60246 Potassium formate, BioUltra, ≥99.0% (NT)
CHKO2
247596
Sodium formate, ACS reagent, ≥99.0%
CHNaO2
107603
Sodium formate, reagent grade, 97%
CHNaO2
456020
Sodium formate, 99.998% trace metals basis
CHNaO2
279412
Sodium formate-13C, 99 atom % 13C
13CHNaO2
71800
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
C2Na2O4
223433
Sodium oxalate, ACS reagent, ≥99.5%
C2Na2O4
71804
Sodium oxalate, certified reference material for titrimetry, certified by BAM, ≥99.5%
C2Na2O4