EMAIL THIS PAGE TO A FRIEND
Journal of microbiology (Seoul, Korea)
Screening of mutant strain Streptomyces mediolani sp. AC37 for (-)-8-O-methyltetrangomycin production enhancement.
PMID 23274989
Abstract
Streptomyces mediolani sp. AC37 was isolated from the root system of higher plant Taxus baccata and produced metabolite identified as (-)-8-O-methyltetrangomycin according to LC/MS/MS analysis. In our screening program for improvements of bioactive secondary metabolites from plant associate streptomycetes, mutation was used as a tool for the induction of genetic variations for selection of higher (-)-8-O-methyltetrangomycin producers of isolates. S. mediolani sp. AC37 was treated with UV irradiation and chemical mutagenic treatment (N-nitroso-N-methyl-urea). The radical scavenging and antioxidant capacity of (-)-8-O-methyltetrangomycin and extracts isolated from mutants were tested using EPR spin trapping technique and ABTS(·+) assay. Comparison of electron microscopic images of Streptomyces sp. AC37 and mutant strains of Streptomyces sp. AC37 revealed substantial differences in morphology and ultrastructure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00034
6-Deoxy-8-O-methylrabelomycin, ≥95% (LC/MS-ELSD)
C20H16O5