EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Antinociceptive effect of Lecythis pisonis Camb. (Lecythidaceae) in models of acute pain in mice.
PMID 23276784
Abstract
Lecythis pisonis Camb., also known in Brazil as sapucaia, is used in folk medicine against pruritus, muscle pain and gastric ulcer. To investigate the antinociceptive effect of ethanol extract from Lecythis pisonis leaves (LPEE), fractions (hexane-LPHF, ether-LPEF and ethyl acetate-LPEAF) and mixture of triterpenes [ursolic and oleanolic acids (MT)] in mice. LPEE and LPEF were evaluated on the acetic acid induced writhings and formalin, capsaicin and glutamate tests. In addition, MT was investigated on the writhings induced by acetic acid, capsaicin and glutamate tests. In the study of some possible mechanisms involved on the antinociceptive effect of LPEF, it was investigated the participation of opioid system, K+ATP channels and L-arginine-nitric oxide pathway. LPEE (12.5 and 25 mg/kg, p.o.), LPEF and MT (6.25, 12.5 and 25 mg/kg, p.o.) reduced the writhings in comparison to saline. LPEE (100 mg/kg, p.o.) and LPEF (50 mg/kg, p.o.) were effective in inhibiting both phases of formalin test. In capsaicin test, LPEE (100 and 200 mg/kg, p.o.), LPEF (12.5-50 mg/kg, p.o) and MT (6.25-25 mg/kg, p.o.) showed a significant antinociceptive effect compared to the control. LPEE (25 and 50 mg/kg, p.o.), LPEF (50 and 100 mg/kg, p.o.) and MT (12.5 and 25 mg/kg, p.o.) reduced the glutamate-evoked nociceptive response. Treatment with naloxone, L-arginine and glibenclamide reversed the effect of LPEF in glutamate test. These results indicate the antinociceptive effect of Lecythis pisonis leaves and suggest that this effect may be related to opioid pathway, K+ATP channels, and L-arginine-nitric oxide modulation. Furthermore, these data support the ethnomedical use of this plant.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0325000
Glybenclamide, European Pharmacopoeia (EP) Reference Standard
C23H28ClN3O5S