EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Suppression of FOXQ1 in benzyl isothiocyanate-mediated inhibition of epithelial-mesenchymal transition in human breast cancer cells.
PMID 23276794
Abstract
We showed previously that breast cancer chemoprevention with benzyl isothiocyanate (BITC) in MMTV-neu mice was associated with induction of E-cadherin protein in vivo. Loss of E-cadherin expression and induction of mesenchymal markers (e.g. vimentin) are biochemical hallmarks of epithelial-mesenchymal transition (EMT), a developmental process implicated in progression of cancer to aggressive state. This study offers novel insights into the mechanism by which BITC inhibits EMT. Exposure of MDA-MB-231, SUM159 and MDA-MB-468 human breast cancer cells to BITC (2.5 and 5 µM) resulted in transcriptional repression of urokinase-type plasminogen activator (uPA) as well as its receptor (uPAR). However, ectopic expression of uPAR in MDA-MB-468 cells failed to confer protection against induction of E-cadherin and inhibition of cell invasion/migration resulting from BITC treatment. The BITC-mediated induction of E-cadherin and inhibition of cell migration was sustained in MDA-MB-231 and SUM159 cells transiently transfected with an uPAR-targeted small interfering RNA. Overexpression of Forkhead Box Q1 (FOXQ1), whose protein and messenger RNA levels were decreased by BITC treatment in cells and MDA-MB-231 xenografts, conferred marked protection against BITC-mediated inhibition of EMT and cell migration. In conclusion, this study implicates FOXQ1 suppression in BITC-mediated inhibition of EMT in human breast cancer cells.