EMAIL THIS PAGE TO A FRIEND
Frontiers in bioscience (Landmark edition)
Nutrition in oncologic patients during antiblastic treatment.
PMID 23276913
Abstract
Cancer may induce weight loss and cachexia, and cancer treatment may contribute to nutritional impairment. Here, we review the literature on the mechanisms of cancer cachexia and the pharmacological interventions both in use in clinical practice and currently under development. Based on this analysis, several nutritional proposals for cancer patients are suggested and the importance of good nutritional status in candidates for hematopoietic stem cell transplantation is highlighted.