EMAIL THIS PAGE TO A FRIEND
Annals of internal medicine
Fatal exserohilum meningitis and central nervous system vasculitis after cervical epidural methylprednisolone injection.
PMID 23277893
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1755000 Methylprednisolone acetate
C24H32O6