EMAIL THIS PAGE TO A FRIEND
Veterinary clinical pathology
Cestode infection in 2 dogs: cytologic findings in liver and a mesenteric lymph node.
PMID 23278428
Abstract
Mesocestoides cestode infections in dogs are well known for causing severe peritonitis with larvae or larval fragments (metacestodes, tetrathyridia, or calcareous corpuscles) frequently observed cytologically in peritoneal fluid samples. This case report describes the cytologic and clinical features of 2 dogs infected with cestode larvae, with one case confirmed and the other presumed to be Mesocestoides sp. In these 2 unusual cases, cestode larvae or larval fragments were found in fine-needle aspirates of the liver and a mesenteric lymph node, but no organisms were found in peritoneal fluid samples. The data presented in this report indicate that clinical pathologists should not rule out Mesocestoides sp cestodiasis based on the absence of larvae in peritoneal fluid samples from dogs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5396
Fenbendazole, ≥98%
C15H13N3O2S
35032
Fenbendazole, VETRANAL, analytical standard
C15H13N3O2S