EMAIL THIS PAGE TO A FRIEND
The Journal of steroid biochemistry and molecular biology
Allopregnanolone-mediated protective effects of progesterone on tributyltin-induced neuronal injury in rat hippocampal slices.
PMID 23280249
Abstract
Increasing evidence shows that progesterone, a neuroactive steroid, has protective actions in central nervous system, but there is little evidence to show the protective mechanism of progesterone on neurotoxicity induced by environmental chemicals. In this study, we examined the effects of progesterone on neuronal injury induced by tributyltin (TBT) in rat hippocampal slices. Treatment with progesterone dose-dependently suppressed hippocampal neuronal injury induced by TBT. The neuroprotective action of progesterone was completely canceled with pretreatment by finasteride, a 5α-reductase inhibitor, but it was not affected by mifepristone, a progesterone receptor antagonist, or by SU-10603, a cytochrome P450 17α inhibitor. The content of allopregnanolone in the slices was significantly increased by treatment with progesterone, and this increment was greatly suppressed with a pretreatment of finasteride. Treatment with allopregnanolone attenuated neuronal injury induced by TBT in a dose-dependent manner. The neuroprotective effects not only of progesterone but also of allopregnanolone were canceled by bicuculline, a potent gamma-aminobutyric acid A (GABAA) receptor antagonist. Pretreatment with muscimol, a GABAA receptor agonist, attenuated hippocampal neuronal injury elicited by TBT. Taken together, allopregnanolone converted from progesterone in hippocampal slices could protect neurons from TBT-induced neurotoxicity due to a GABAA receptor-dependent mechanism. One of the physiological roles of neuroactive steroids might be neuroprotection from environmental chemicals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8129
5β-Pregnan-3α-ol-20-one
C21H34O2
34202
Finasteride, VETRANAL®, analytical standard
C23H36N2O2