EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Novel quality by design tools for concentrated drug suspensions: surface energy profiling and the fractal concept of flocculation.
PMID 23280339
Abstract
Quality by design is an important concept, but only limited research has been invested in concentrated pharmaceutical suspensions. A need exists for novel analytical tools to thoroughly characterize the drug as well as its aggregated particle structure in suspension. This work focuses on lipid-based pharmaceutical suspensions for filling of capsules. A rheological approach, namely the fractal concept of flocculation, is introduced to the pharmaceutical field. The model drug mebeverine hydrochloride was first physicochemically analyzed. A special aim was to study the surface energy profiles using inverse gas chromatography as a critical characteristic for the suspension's rheological behavior. Suspensions were manufactured in laboratory process equipment while applying different homogenization speeds. Flow curves of the final suspensions were measured using a cone-and-plate rheometer. As a result, surface energy profiles revealed differences from one mebeverine lot to another. Different homogenization intensities greatly affected the viscosity and the Mooney model was able to predict experimental values as a function of the drug volume fraction. The fractal concept of flocculation characterized mebeverine in suspension and a slight increase of fractal dimension was noted when homogenization speed was increased. It was concluded that the introduced concepts have large potential for designing quality into concentrated pharmaceutical suspensions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3282
Mebeverine hydrochloride, analytical standard
C25H35NO5 · HCl