EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
New structural motifs in lithium and zinc calix[4]arene chemistry.
PMID 23280484
Abstract
The structures of three novel compounds prepared from the reaction of the lower-rim 1,3-diacid calix[4]arene (LH(2)) with lithium carbonate (1), t-butyl-lithium (2) and zinc acetate (3) are reported.