EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Hole-transporting transistors and circuits based on the transparent inorganic semiconductor copper(I) thiocyanate (CuSCN) processed from solution at room temperature.
PMID 23280854
Abstract
The wide bandgap and highly transparent inorganic compound copper(I) thiocyanate (CuSCN) is used for the first time to fabricate p-type thin-film transistors processed from solution at room temperature. By combining CuSCN with the high-k relaxor ferroelectric polymeric dielectric P(VDF-TrFE-CFE), we demonstrate low-voltage transistors with hole mobilities on the order of 0.1 cm(2) V(-1) s(-1) . By integrating two CuSCN transistors, unipolar logic NOT gates are also demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
298212
Copper(I) thiocyanate, 99%
CCuNS