EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
SnAP reagents for the transformation of aldehydes into substituted thiomorpholines--an alternative to cross-coupling with saturated heterocycles.
PMID 23281159