EMAIL THIS PAGE TO A FRIEND
Macromolecular rapid communications
Nematic liquid crystalline elastomer grating and microwire fabricated by micro-molding in capillaries.
PMID 23281167
Abstract
A new approach is developed to fabricate highly oriented mono-domain LCE nano/microstructures through micro-molding in capillaries. Gratings and microwires as two typical examples are fabricated and characterized by polarizing optical microscopy, optical microscopy, and scanning electron microscopy. The gratings with precisely controlled sizes and smooth surface are obtained by filling the channels with a nematic monomer mixture followed by the photo-crosslinking. After peeling off the gratings from the substrate, the free-standing microwires are obtained. A uniform orientation of the mesogenic units is observed for the molds with channel width less than 20 μm. Reversible thermomechanical effect is demonstrated by using the microwires obtained through this approach.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
290009
Potassium benzoate, ReagentPlus®, 99%
C7H5KO2