EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Paeoniflorin acts as a liver X receptor agonist.
PMID 23281636
Abstract
Paeoniflorin is one of the active ingredients of Paeonia lactiflora Pall., a novel traditional herbal medicine exerting pharmacological effects including antihyperlipidemic, neuroprotective, and anti-hepatofibrosis effects. Liver X receptor (LXR) acts as a ligand-activated transcription factor to exhibit antihyperlipidemic and neuroprotective effects. In this study, the activity of paeoniflorin against LXR was evaluated by the mammalian one-hybrid and transient transfection reporter assays. The results showed that paeoniflorin transactivated GAL4, rat cholesterol 7 α-hydroxylase, phospholipid transfer protein, and ATP-binding cassette A1 gene promoters in dose-dependent manner. Furthermore, the docking study demonstrated that paeoniflorin resided in the LXR ligand-binding pocket in the similar manner as GSK 3987, a novel LXR agonist. These results indicated that paeoniflorin might exert pharmacological effects through LXR pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0038
Paeoniflorin, ≥98% (HPLC)
C23H28O11