EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Cyclodextrin-modified micellar electrokinetic chromatography for enantioseparations.
PMID 23283789
Abstract
The separation of enantiomers is one of the important fields of modern analytical chemistry, especially for agrochemical and pharmaceutical products because the stereochemistry has a significant influence on the biological activities of compounds. Cyclodextrin-modified micellar electrokinetic chromatography (CD-MEKC) has become an important capillary electrophoresis mode for enantioseparations. Here, we describe an example of a CD-MEKC method using hydroxypropyl-γ-cyclodextrin as chiral selector and sodium dodecyl sulfate as micellar solution for enantioseparation of triazole fungicides and the drug econazole.