PloS one
iNOS activation regulates β-catenin association with its partners in endothelial cells.
PMID 23285236
Abstract
Signals that disrupt β-catenin association to cadherins may influence the translocation of β-catenin to the nucleus to regulate transcription. Post-translational modification of proteins is a signalling event that may lead to changes in structural conformation, association or function of the target proteins. NO and its derivatives induce nitration of proteins during inflammation. It has been described that animals treated with NO donors showed increased permeability due to modulation of VE-cadherin/catenin complex. We, therefore, aim to evaluate the effect of iNOS activation on the expression, nuclear localisation and function of β-catenin in endothelial cells. Expression, nuclear localisation, post-translational modifications and function of β-catenin was analysed by cell fractionation, immunoprecipitation, immunoblots, QRT-PCR and permeability assays in murine endothelial cells (H5V). Influence of macrophage activation on expression of VE-cadherin/p120-catenin/β-catenin complex in co-cultured H5V cells was also assessed. Activation of macrophages to produce NO provoked a decrease in VE-cadherin/p120-catenin/β-catenin expression in H5V cells. Phosphorylation of β-catenin, p120-catenin and VE-cadherin, and reduction in the barrier properties of the cell monolayer was associated with iNOS induction. Moreover, high NO levels provoked nitration of β-catenin, and induced its translocation to the nucleus. In the nucleus of NOS activated cells, nitration levels of β-catenin influenced its association with TCF4 and p65 proteins. High levels of NO altered β-catenin mediated gene expression of NFκB and Wnt target genes without affecting cell viability. NOS activity modulates β-catenin post-translational modifications, function and its association with different partners to promote endothelial cell survival. Therapeutic manipulation of iNOS levels may remove a critical cytoprotective mechanism of importance in tumour angiogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01423
N-Acetyl-D-penicillamine, for HPLC derivatization, ≥99.0% (T)
C7H13NO3S