EMAIL THIS PAGE TO A FRIEND
Comparative biochemistry and physiology. B, Comparative biochemistry
Structural stability of fish myosin subfragment-1.
PMID 2328565
Abstract
1. Tryptic cleavage of fish myosin subfragment-1 (S-1) revealed its similar substructure of heavy chain to that of rabbit S-1. 2. The structural stability of fish S-1 was studied by thermal denaturation method, and a rapid polymerization of inactivated fish S-1, detected by turbidity increase, was characteristic. 3. The light-chain release and tryptic susceptibility increase upon heating were significant with fish S-1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CL4B200 Sepharose® CL-4B, Cross-linked