EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Clinical re-evaluation of effects of two different "cocktail therapy" to prevent from phlebitis induced by Chansu injection].
PMID 23285923
Abstract
To re-evaluate the effects of different "cocktail therapy" to prevent from phlebitis induced by Chansu injection. Patients treated with Chansu injection were divided randomLy into 4 groups with 90 per group, control group, phentolaminum group, the magnesium sulfate group-phentolaminum group, and anisodamine-phentolaminum group. Patients in the control group only received the routine nursing treatment, and patients in the various experiment group received different interventions. The comparison was made in the morbidity and the starting time of occurrence of phlebitis, the severity of pain, duration of pain. The morbidity of phlebitis was 8%, 8%, 6%, respectively. The starting time of phlebitis occurrence was (22 +/- 4), (27 +/- 5), (28 +/- 7) h, respectively. The NRS of pain was (4.75 +/- 1.51), (3.27 +/- 1.02), (2.71 +/- 1.63), respectively. The duration time of pain was (4.25 +/- 1.36), (2.51 +/- 1.05), (2.19 +/- 1.13) d respectively. In control group, the morbidity of phlebitis, the starting time of occurrence of phlebitis, the severity of pain, duration of pain was 30%, (16 +/- 4) h, (6.34 +/- 1.21), (5.47 +/- 1.07) d, respectively. As compared with the control group, a significance difference was found between every group in three test groups and control group respectively (P<0.05). The morbidity and the starting time of occurrence of phlebitis, the severity of pain, duration of pain was significantly reduced respectively by two different "cocktail therapy".
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7547
Phentolamine hydrochloride, ≥98% (TLC), powder
C17H19N3O · HCl
P7561
Phentolamine methanesulfonate salt, ≥98% (TLC), powder
C17H19N3O · CH4O3S