EMAIL THIS PAGE TO A FRIEND
Immunohematology
Warm autoadsorption with enzyme-treated red blood cells.
PMID 23286554
Abstract
Patients demonstrating warm autoantibody specificity present serologic challenges for laboratory staff performing antibody identification in the blood bank. Autoantibody can be removed from plasma or serum by adsorption onto autologous red blood cells (RBCs) provided the patient has not been transfused in the previous 3 months. The adsorption process can be enhanced by enzyme pretreatment of autologous RBCs.