EMAIL THIS PAGE TO A FRIEND
Journal of cosmetic science
Protection against UVB-induced oxidative stress in human skin cells and skin models by methionine sulfoxide reductase A.
PMID 23286868
Abstract
Environmental trauma to human skin can lead to oxidative damage of proteins and affect their activity and structure. When methionine becomes oxidized to its sulfoxide form, methionine sulfoxide reductase A (MSRA) reduces it back to methionine. We report here the increase in MSRA in normal human epidermal keratinocytes (NHEK) after ultraviolet B (UVB) radiation, as well as the reduction in hydrogen peroxide levels in NHEK pre-treated with MSRA after exposure. Further, when NHEK were pre-treated with a non-cytotoxic pentapeptide containing methionine sulfoxide (metSO), MSRA expression increased by 18.2%. Additionally, when the media of skin models were supplemented with the metSO pentapeptide and then exposed to UVB, a 31.1% reduction in sunburn cells was evident. We conclude that the presence of MSRA or an externally applied peptide reduces oxidative damage in NHEK and skin models and that MSRA contributes to the protection of proteins against UVB-induced damage in skin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64430
DL-Methionine sulfoxide, ≥99.0% (NT)
C5H11NO3S
M1126
L-Methionine sulfoxide
C5H11NO3S