EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Discovery of coumarin-monastrol hybrid as potential antibreast tumor-specific agent.
PMID 23287057
Abstract
Development of new, targeted antibreast cancer drug which can treat both the hormone receptor (positive and negative) breast cancers is a very challenging task. The concept of molecular hybridization led us to discover a novel class of coumarin-monastrol hybrid, as a novel breast cancer agent which selectively induce apoptosis in both primary and metastatic breast cancer cell lines.