EMAIL THIS PAGE TO A FRIEND
Journal of clinical psychopharmacology
Ifenprodil for emotional incontinence in patients with vascular dementia: two case reports.
PMID 23288243
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2892
Ifenprodil (+)-tartrate salt
C42H54N2O4 · C4H6O6