EMAIL THIS PAGE TO A FRIEND
Molecular diversity
A mild and efficient one-pot synthesis of β-amino carbonyl compounds via Mannich reaction under ambient temperature condition.
PMID 23288546
Abstract
We have developed an efficient protocol for the synthesis of β-amino carbonyl compounds using phenylboronic acid as effective and eco-friendly catalyst via one-pot three-component Mannich reaction. Excellent yields, mild reaction condition, and simple experimental work-up procedure are some of the advantages of this method, which makes it a useful protocol for the synthesis of β-amino carbonyl compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78181
Phenylboronic acid, purum, ≥97.0% (HPLC)
C6H7BO2