EMAIL THIS PAGE TO A FRIEND
Doklady. Biochemistry and biophysics
Benzophenone derivatives of pyrimidines as effective non-nucleoside inhibitors of wild-type and drug-resistant HIV-1 reverse transcriptase.
PMID 23288568