EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Ruthenium-triggered ring opening of ethynylcyclopropanes: [3+2] cycloaddition with aldehydes and aldimines involving metal allenylidene intermediates.
PMID 23288721
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295183
Cyclopropane, ≥99%
C3H6
185922
Furan, ≥99%
C4H4O
43861
Furan, analytical standard
C4H4O