EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Effect of the pesticide tocuthion on the activity of microsomal cytochrome P-450-dependent monooxygenases in the rat liver].
PMID 2328924
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45311
Prothiofos, PESTANAL®, analytical standard
C11H15Cl2O2PS2