EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Oxidation of aromatic N-heterocyclic compounds to N-oxides by Verticillium sp. GF39 cells.
PMID 23290448
Abstract
Verticillium sp. GF39, catalyzing the oxidation of 1-methylisoquinoline to 1-methylisoquinoline N-oxide, was found to be the highest N-oxide producer. Under the optimized reaction conditions, the whole cells of Verticillium sp. GF39 formed 5 mM 1-methylisoquinoline N-oxide from 1-methylisoquinoline with a molar conversion yield of 100% after a 10-h incubation at 20°C. The whole cells also acted on pyridine, 2-methylpyridine, quinoline and isoquinoline and formed the corresponding N-oxides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
264938
1-Methylisoquinoline, 97%
C10H9N