EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Dissolution of tablet-in-tablet formulations studied with ATR-FTIR spectroscopic imaging.
PMID 23291036
Abstract
This work uses ATR-FTIR spectroscopic imaging to study the dissolution of delayed release and pH resistant compressed coating pharmaceutical tablets. Tablets with an inner core and outer shell were constructed using a custom designed compaction cell. The core of the delayed release tablets consisted of hydroxypropyl methylcellulose (HPMC) and caffeine. The shell consisted of microcrystalline cellulose (MCC) and glucose. The core of the pH resistant formulations was an ibuprofen and PEG melt and the shell was constructed from HPMC and a basic buffer. UV/vis spectroscopy was used to monitor the lag-time of drug release and visible optical video imaging was used as a complementary imaging technique with a larger field of view. Two delayed release mechanisms were established. For tablets with soluble shell sections, lag-time was dependent upon rapid shell dissolution. For tablets with less soluble shells, the lag-time was controlled by the rate of dissolution medium ingress through the shell and the subsequent expansion of the wet HPMC core. The pH resistant formulations prevented crystallization of the ibuprofen in the core during dissolution despite an acidic dissolution medium. FTIR imaging produced important information about the physical and chemical processes occurring at the interface between tablet sections during dissolution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7509
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
H9262
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
H8384
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
09963
(Hydroxypropyl)methyl cellulose
423238
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
423203
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
423181
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
423173
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
H3785
Hypromellose, meets USP testing specifications
64670
Methocel® 65 HG, 27-29% methoxyl basis
64680
Methocel® 90 HG, 22-27% methoxyl basis
64605
Methocel® A15 LV, 27.5-31.5% methoxyl basis
64625
Methocel® A15C, medium viscosity, Methoxyl content 27.5-31.5 %
94378
Methocel® A4M, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
M0262
Methyl cellulose, viscosity: 400 cP
M7140
Methyl cellulose, viscosity: 15 cP
M6385
Methyl cellulose, viscosity: 25 cP
M7027
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
M0387
Methyl cellulose, viscosity: 1,500 cP
M0430
Methyl cellulose, meets USP testing specifications, viscosity: 400 cP
M0555
Methyl cellulose, meets USP testing specifications, viscosity: 1,500 cP
64632
Methyl cellulose, tested according to Ph Eur