EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and structure-activity relationships of harmine derivatives as potential antitumor agents.
PMID 23291116
Abstract
Harmine, a naturally occurring β-carboline alkaloid, showed good antitumor activities together with remarkable neurotoxic effects in animal models. In order to search for novel leading compounds endowed with better antitumor activities and less neurotoxicities, a series of harmine derivatives were designed and synthesized by modification of position-2, 7 and 9 of β-carboline nucleus, and their cytotoxic activities against human tumor cell lines were investigated. Acute toxicities and antitumor activities of the selected compounds in mice were also evaluated. Structure-activity relationships studies confirmed that (1) the 7-methoxy structural moiety was the pharmacophore responsible for the neurotoxic effects of this class of compounds; (2) the substituents in position-2 and 9 played a vital role in modulation of their antitumor activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
286044
Harmine, 98%
C13H12N2O