EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Enzymatic improvement in the polyphenol extractability and antioxidant activity of green tea extracts.
PMID 23291774
Abstract
This study describes increases in extraction efficiency and the bioconversion of catechins after treatment with several commercial enzymes. Tannase was also used to improve the anti-radical activities of green tea extracts. Enzymatic treatment with various commercial enzymes was introduced to improve the extraction efficiency of polyphenols. The total polyphenol, flavonoid, and catechin contents and the 1,1-diphenyl-2-picrylhydrazyl (DPPH) radical scavenging activity of the green tea extract treated with Viscozyme (VG) were significantly higher than those treated with other commercial enzymatic extractions (p<0.05). More than 95% of the epigallocatechingallate (EGCG) and of the epicatechingallate (ECG) was hydrolyzed to epigallocatechin (EGC) and to epicatechin (EC) in successive 20 min treatments with Viscozyme and tannase (TG). Due to its hydrolytic activity, treatment involving tannase resulted in a significant release of gallic acid (GA), EGC, and EC, leading to greater radical scavenging activities. Regarding the IC(50) values of the DPPH and 2,2-azino-di-(3-ethylbenzothiazoline-6-sulfonic acid) (ABTS) radicals, the green tea extract treated with TG showed values of 131.23 and 28.83 µg/mL, VG showed values of 224.70 and 32.54 µg/mL, and normal green tea extract (NG) showed values of 241.11 and 66.27 µg/mL, respectively. These results indicate that successive treatment with Viscozyme and tannase improves the extraction efficiency of polyphenols and increases radical scavenging activities.