EMAIL THIS PAGE TO A FRIEND
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova
[Investigation and analysis of chloride channels distribution over the surface and T-tubule membranes of frog skeletal muscle].
PMID 23293819
Abstract
Two types of muscle fibre integral currents (T1T and T2T) were recorded using narrow-tipped extracellular pipettes in isolated sartorius muscles of frog, Rana temporaria. T1T and T2T responses presumably were generated by currents originating in the muscle fibers sarcolemma (M-band region) or both sarcolemma and T-system (Z-line region), respectively, and differently responded to the selective blockade of C1C chloride channels with anthracene-9-carboxylic acid (9-AC). Eva- luation of the role of prolongation of T1T responses in generation of multiple peaks of the second phase (Na current) of T2T integral currents in muscle fibers are discussed.
Related Materials