EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
Copper-zinc superoxide dismutase-deficient mice show increased susceptibility to experimental autoimmune encephalomyelitis induced with myelin oligodendrocyte glycoprotein 35-55.
PMID 23294897
Abstract
In this report, we have addressed the role of copper-zinc superoxide dismutase (SOD1) deficiency in the mediation of central nervous system autoimmunity. We demonstrate that SOD1-deficient C57Bl/6 mice develop more severe autoimmune encephalomyelitis induced with myelin oligodendrocyte glycoprotein (MOG) 35-55, compared with wild type mice. This alteration in the disease phenotype was not due to aberrant expansion of MOG-specific T cells nor their ability to produce inflammatory cytokines; rather lymphocytes generated in SOD1-deficient mice were more prone to spontaneous cell death when compared with their wild type littermate controls. The data point to a role for SOD1 in the maintenance of self-tolerance leading to the suppression of autoimmune responses.