EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Mechanisms of the thermal decay of chlorpropham.
PMID 23295769
Abstract
DFT calculations were performed on the thermal reactions of chlorpropham 1, a carbamate pesticide and plant growth regulator frequently used in the storage of potatoes. At the conditions normally used in applying 1 (injection of a methanolic solution of 1 into a hot air stream, T ≈ 500°C), both ester pyrolysis of 1 and a methanol-or water-catalysed isocyanate cleavage are expected to proceed rapidly (lifetime of 1 less than a second). In both reactions, the final reaction product will be toxic and carcinogenic m-chloroaniline 2. Matrix-isolation experiments indicate that 1 undergoes thermal decay at temperatures as low as 250°C. Up to temperatures of ca. 500°C, formation of m-chlorophenylisocyanate 4 and isopropanol was the predominant reaction observed, while formation of propene, CO(2), and m-chloroaniline 2 was the most important reaction channel at higher pyrolysis temperatures. m-Chlorophenyl carbamic acid 3 could not be observed. The results indicate that at lower temperatures, 1 decays exclusively via isocyanate cleavage of 1, provided that traces of catalytic water or other protic compounds are present. At higher temperatures, ester cleavage of 1 becomes competitive and outweighs the isocyanate cleavage by a factor of ca. 10:1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45393
Chlorpropham, PESTANAL®, analytical standard
C10H12ClNO2