EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Ganglioside-embedding small bicelles for probing membrane-landing processes of intrinsically disordered proteins.
PMID 23296043
Abstract
We successfully displayed a series of gangliosides on small bicelles with a uniform confined size, offering nanoscale standardized membrane mimics for spectroscopic characterization of weak encounter complexes formed between ganglioside clusters and amyloidogenic proteins. This enabled probing of initial membrane-landing processes of α-synuclein as a therapeutic target by NMR spectroscopy.