EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Characterization of membrane-associated glycoproteins using lectin affinity chromatography and mass spectrometry.
PMID 23296525
Abstract
Membrane-associated glycoproteins play critical roles in many biological processes and are often the therapeutic targets for drug discovery. Lectin affinity chromatography is one of the most widely used approaches for enrichment of glycoproteins at the protein level. Here, we describe a strategy for the characterization of membrane glycoproteins including membrane protein extraction, lectin affinity chromatography, protein digestion, and analysis by LC-MS/MS.