EMAIL THIS PAGE TO A FRIEND
Osteoporosis international : a journal established as result of cooperation between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA
Promotion of peri-implant bone healing by systemically administered parathyroid hormone (1-34) and zoledronic acid adsorbed onto the implant surface.
PMID 23296744
Abstract
The effect of human parathyroid hormone 1-34 (PTH) and zoledronic acid (ZA) alone or in combination on bone healing in osteoporotic settings was tested using implants inserted in tibiae of ovariectomized (OVX) rats. Combination therapy promoted bone healing more than each treatment alone 12 weeks after implant insertion. PTH and ZA have been demonstrated to be effective on implant fixation. However, reports about the combined use of PTH and ZA for promotion of bone healing around implant in osteoporotic settings are still limited. This study aims to investigate effects of PTH+ZA on implant stabilization in OVX rats. Twelve weeks after bilateral ovariectomy, OVX rats randomly received implants without or with ZA (by immersion in 1 mg/ml ZA solution for 24 h). Subsequently, half of the animals from each group also received subcutaneous injections of PTH (60 μg/kg, three times a week) for 12 weeks. Thus, there were four groups: control, PTH, ZA, and PTH+ZA. All treatments promoted bone healing around implant compared to control, but PTH+ZA treatment showed significantly stronger effects than PTH or ZA alone in histological, micro-CT, and biomechanical tests. The results indicated the additive effects of PTH and ZA on implant fixation in OVX rats; it was suggested that the anabolic effect of PTH was potent and not blunted by ZA during bone healing around implant when used concurrently.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3796 Parathyroid Hormone Fragment 1-34 human, ≥95% (HPLC), powder
C181H291N55O51S2