EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
The structural plasticity of the proximal [4Fe3S] cluster is responsible for the O2 tolerance of membrane-bound [NiFe] hydrogenases.
PMID 23296978