EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Using a two-stage hydrogen-based membrane biofilm reactor (MBfR) to achieve complete perchlorate reduction in the presence of nitrate and sulfate.
PMID 23298383
Abstract
We evaluated a strategy for achieving complete reduction of perchlorate (ClO(4)(-)) in the presence of much higher concentrations of sulfate (SO(4)(2-)) and nitrate (NO(3)(-)) in a hydrogen-based membrane biofilm reactor (MBfR). Full ClO(4)(-) reduction was achieved by using a two-stage MBfR with controlled NO(3)(-) surface loadings to each stage. With an equivalent NO(3)(-) surface loading larger than 0.65 ± 0.04 g N/m(2)-day, the lead MBfR removed about 87 ± 4% of NO(3)(-) and 30 ± 8% of ClO(4)(-). This decreased the equivalent surface loading of NO(3)(-) to 0.34 ± 0.04-0.53 ± 0.03 g N/m(2)-day for the lag MBfR, in which ClO(4)(-) was reduced to nondetectable. SO(4)(2-) reduction was eliminated without compromising full ClO(4)(-) reduction using a higher flow rate that gave an equivalent NO(3)(-) surface loading of 0.94 ± 0.05 g N/m(2)-day in the lead MBfR and 0.53 ± 0.03 g N/m(2)-day in the lag MBfR. Results from qPCR and pyrosequencing showed that the lead and lag MBfRs had distinctly different microbial communities when SO(4)(2-) reduction took place. Denitrifying bacteria (DB), quantified using the nirS and nirK genes, dominated the biofilm in the lead MBfR, but perchlorate-reducing bacteria (PRB), quantified using the pcrA gene, became more important in the lag MBfR. The facultative anaerobic bacteria Dechloromonas, Rubrivivax, and Enterobacter were dominant genera in the lead MBfR, where their main function was to reduce NO(3)(-). With a small NO(3)(-) surface loading and full ClO(4)(-) reduction, the dominant genera shifted to ClO(4)(-)-reducing bacteria Sphaerotilus, Rhodocyclaceae, and Rhodobacter in the lag MBfR.