EMAIL THIS PAGE TO A FRIEND
Veterinary parasitology
Prevention of transmission of Ehrlichia canis by Rhipicephalus sanguineus ticks to dogs treated with a combination of fipronil, amitraz and (S)-methoprene (CERTIFECT®).
PMID 23298566
Abstract
The ability of CERTIFECT(®) (a combination of fipronil, amitraz and (S)-methoprene) to prevent transmission of Ehrlichia canis was studied in two groups of eight dogs. One group was treated with CERTIFECT while the other group remained untreated. All dogs were exposed to E. canis-infected Rhipicephalus sanguineus ticks on Days 7, 14, 21 and again on day 28 post-treatment by releasing ticks into the kennels of the dogs to simulate the natural way of infestation. Animals were examined in situ for ticks on Days 9, 16 and 23 and any ticks present were counted and removed on Day 30. The efficacy of CERTIFECT against R. sanguineus was 100%, since no ticks were found on the treated dogs at any time. Clinical examinations (including monitoring body temperature and blood collections for PCR analysis and serology) were performed at intervals throughout the study until Day 56. Five out of 8 untreated control dogs (62.5%) became infected with E. canis, as demonstrated by detection of specific E. canis antibodies and the presence of E. canis DNA in blood samples by PCR. These dogs displayed fever and thrombocytopenia and were rescue-treated with doxycline. None of the 8 dogs treated with CERTIFECT became infected with E. canis, in comparison to the 5/8 control dogs, as confirmed by the lack of specific antibodies and absence of any ehrlichial DNA in blood samples by PCR. CERTIFECT prevented transmission of E. canis and effectively provided protection against monocytic ehrlichiose for at least 4 weeks post treatment.
Related Materials