EMAIL THIS PAGE TO A FRIEND
Nanoscale
A conceptual translation of homogeneous catalysis into heterogeneous catalysis: homogeneous-like heterogeneous gold nanoparticle catalyst induced by ceria supporter.
PMID 23299453
Abstract
Translation of homogeneous catalysis into heterogeneous catalysis is a promising solution to green and sustainable development in chemical industry. For this purpose, noble metal nanoparticles represent a new frontier in catalytic transformations. Many challenges remain for researchers to transform noble metal nanoparticles of heterogeneous catalytic active sites into ionic species of homogeneous catalytic active sites. We report here a successful design on translating homogeneous gold catalysis into a heterogeneous system with a clear understanding of the catalytic pathway. This study initiates a novel concept to immobilize a homogeneous catalyst based on electron transfer between supporting base and supported nanoparticles. Meanwhile, on the basis of theoretical calculation, it has deepened the understanding of the interactions between noble metal nanoparticles and the catalyst support.