EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Corrections for "Thermodynamic modeling of activity coefficient and prediction of solubility. Parts 1 and 2," Journal of Pharmaceutical Sciences 95(4) 2006, pp 790-809.
PMID 23299567