EMAIL THIS PAGE TO A FRIEND
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery
β-Tricalcium phosphate implant to repair empty nose syndrome: preliminary results.
PMID 23300225