EMAIL THIS PAGE TO A FRIEND
Scientific reports
Discovery of highly potent acid ceramidase inhibitors with in vitro tumor chemosensitizing activity.
PMID 23301156
Abstract
The expression of acid ceramidase (AC) - a cysteine amidase that hydrolyses the proapoptotic lipid ceramide - is abnormally high in several human tumors, which is suggestive of a role in chemoresistance. Available AC inhibitors lack, however, the potency and drug-likeness necessary to test this idea. Here we show that the antineoplastic drug carmofur, which is used in the clinic to treat colorectal cancers, is a potent AC inhibitor and that this property is essential to its anti-proliferative effects. Modifications in the chemical scaffold of carmofur yield new AC inhibitors that act synergistically with standard antitumoral drugs to prevent cancer cell proliferation. These findings identify AC as an unexpected target for carmofur, and suggest that this molecule can be used as starting point for the design of novel chemosensitizing agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1494
Carmofur, ≥98% (HPLC), powder
C11H16FN3O3